N1-(5-Chloropyridin-2-yl)-N2-((1S,2R,4S)-4-(dimethylcarbamoyl)-2-(5-methyl-4,5,6,7-tetrahydrothiazolo[5,4-c]pyridine-2-carboxamido)cyclohexyl)oxalamide 98%

Supplier: Ambeed
Danger

Synonyms: Edoxaban

A587736-100MG A587736-10MG A587736-1G A587736-50MG A587736-250MG A587736-5G A587736-25G A587736-5MG
76819-302EA 10.61 USD
76819-302 76819-304 76819-306 76819-308 77594-784 77613-820 77673-580 77673-582
N1-(5-Chloropyridin-2-yl)-N2-((1S,2R,4S)-4-(dimethylcarbamoyl)-2-(5-methyl-4,5,6,7-tetrahydrothiazolo[5,4-c]pyridine-2-carboxamido)cyclohexyl)oxalamide 98%
N1-(5-Chloropyridin-2-yl)-N2-((1S,2R,4S)-4-(dimethylcarbamoyl)-2-(5-methyl-4,5,6,7-tetrahydrothiazolo[5,4-c]pyridine-2-carboxamido)cyclohexyl)oxalamide

Precaución: for Research Use Only

Formula: C₂₄H₃₀ClN₇O₄S
MW: 548.06 g/mol
MDL Number: MFCD13195544
CAS Number: 480449-70-5
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR