N-Benzyl 4-boronobenzenesulfonamide 98%

Supplier: Ambeed
Warning

Synonyms: 4-(N-Benzylsulphonamido)benzeneboronic acid, (4-(N-Benzylsulphamoyl)phenyl)boronic acid

A587430-1G A587430-5G
76819-298EA 72.15 USD
76819-298 76819-300
N-Benzyl 4-boronobenzenesulfonamide 98%
N-Benzyl 4-boronobenzenesulphonamide

Precaución: for Research Use Only

Formula: C₁₃H₁₄BNO₄S
MW: 291.1306 g/mol
MDL Number: MFCD06659845
CAS Number: 548769-96-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR