2-Methoxybenzylamine 97%

Supplier: Ambeed
Danger

A587328-10G A587328-1G A587328-25G A587328-5G
76819-292EA 35 USD
76819-292 76819-294 76819-296 77572-868
2-Methoxybenzylamine 97%
2-Methoxybenzylamine

Precaución: for Research Use Only

Formula: C₈H₁₁NO
MDL Number: MFCD00008110
CAS Number: 6850-57-3
UN: 2735
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR