3,5-Dimethylpyridin-2-amine 98%

Supplier: Ambeed
Warning

A585755-100G A585755-10G A585755-1G A585755-250MG A585755-25G A585755-5G
76819-268EA 615 USD
76819-268 76819-270 76819-272 76819-274 76819-276 76819-278
3,5-Dimethylpyridin-2-amine 98%
3,5-Dimethylpyridin-2-amine

Precaución: for Research Use Only

Formula: C₇H₁₀N₂
MDL Number: MFCD11040341
CAS Number: 41995-30-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR