N-Methylpyrrole-3-boronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 1-METHYL-1H-PYRROL-3-YLBORONIC ACID PINACOL ESTER, 1H-Pyrrole,1-methyl-3-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-, 1-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrole

A581232-1G A581232-250MG A581232-5G
76819-260EA 30 USD
76819-260 76819-262 76819-264
N-Methylpyrrole-3-boronic acid pinacol ester 98%
N-Methylpyrrole-3-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₁H₁₈BNO₂
MW: 207.08 g/mol
MDL Number: MFCD13182200
CAS Number: 953040-54-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR