6,7-Dimethoxyquinolin-4(1H)-one 95%

Supplier: Ambeed
Warning

A579010-1G A579010-5G
76819-256EA 6.37 USD
76819-256 76819-258
6,7-Dimethoxyquinolin-4(1H)-one 95%
6,7-Dimethoxyquinolin-4(1H)-one

Precaución: for Research Use Only

Formula: C₁₁H₁₁NO₃
MW: 205.21 g/mol
MDL Number: MFCD09031937
CAS Number: 127285-54-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR