SB239063 99+%

Supplier: Ambeed
Warning

Synonyms: 4-[4-(4-fluorophenyl)-5-(2-methoxypyrimidin-4-yl)imidazol-1-yl]cyclohexan-1-ol, trans-4-[4-(4-fluorophenyl)-5-(2-methoxy-4-pyrimidinyl)-1H-imidazol-1-yl]-cyclohexanol, Rel-(1r,4r)-4-(4-(4-fluorophenyl)-5-(2-methoxypyrimidin-4-yl)-1H-imidazol-1-yl)cyclohexan-1-ol, 4-(4-(4-fluorophenyl)-5-(2-methoxypyrimidin-4-yl)-1H-imidazol-1-yl)cyclohexanol

A577018-10MG A577018-1MG A577018-50MG A577018-5MG
76819-242EA 71.09 USD
76819-242 76819-244 76819-246 76819-248
SB239063 99+%
SB239063

Precaución: for Research Use Only

Formula: C₂₀H₂₁FN₄O₂
MW: 368.4 g/mol
MDL Number: MFCD03792786
CAS Number: 193551-21-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR