2-Bromo-4-fluorophenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (2-Bromo-4-fluorophenyl)boronic acid

A576140-1G A576140-250MG A576140-5G
76819-228EA 32.89 USD
76819-228 76819-230 76819-232
2-Bromo-4-fluorophenylboronic acid 98%
2-Bromo-4-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₅BBrFO₂
MW: 218.82 g/mol
MDL Number: MFCD13195647
CAS Number: 1217501-12-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR