5-Bromoquinoline-8-carboxaldehyde 95%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-8-formyl-1-azanaphthalene, 5-Bromoquinoline-8-carbaldehyde, 5-Bromo-8-formylquinoline

A576042-100MG A576042-1G A576042-5G
76819-224EA 41.38 USD
76819-224 76819-226 77673-550
5-Bromoquinoline-8-carboxaldehyde 95%
5-Bromoquinoline-8-carboxaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₆BrNO
MW: 236.065 g/mol
MDL Number: MFCD08064042
CAS Number: 885267-41-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR