3-Carboxy-4-fluorophenylboronic acid pinacol ester 95%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid, 3-Carboxy-4-fluorobenzeneboronic acid pinacol ester

A575885-1G A575885-25G A575885-5G
76819-218EA 22 USD
76819-218 76819-220 76819-222
3-Carboxy-4-fluorophenylboronic acid pinacol ester 95%
3-Carboxy-4-fluorophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₆BFO₄
MDL Number: MFCD12546519
CAS Number: 882679-10-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR