2-Methyl-6-(4-methylpiperazin-1-yl)pyrimidin-4-amine 95%

Supplier: Ambeed
Warning

A575354-250MG A575354-1G
76819-214EA 107.16 USD
76819-214 77673-546
2-Methyl-6-(4-methylpiperazin-1-yl)pyrimidin-4-amine 95%
2-Methyl-6-(4-methylpiperazin-1-yl)pyrimidin-4-amine

Precaución: for Research Use Only

Formula: C₁₀H₁₇N₅
MW: 207.27 g/mol
MDL Number: MFCD21604271
CAS Number: 1104376-15-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR