2-(4-Bromophenoxy)tetrahydropyran 97%

Supplier: Ambeed
Warning

Synonyms: 2-(4-Bromophenoxy)tetrahydro-2H-pyran

A572279-100G A572279-25G A572279-5G
76819-204EA 151.71 USD
76819-204 76819-206 76819-208
2-(4-Bromophenoxy)tetrahydropyran 97%
2-(4-Bromophenoxy)tetrahydropyran

Precaución: for Research Use Only

Formula: C₁₁H₁₃BrO₂
MW: 257.13 g/mol
MDL Number: MFCD00091551
CAS Number: 36603-49-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR