Z-Trp-OBzl 97%

Supplier: Ambeed
Warning

Synonyms: N-Benzyloxycarbonyl-L-tryptophan benzyl ester

A570039-100G A570039-1G A570039-25G
76819-192EA 278.01 USD
76819-192 76819-194 76819-196
Z-Trp-OBzl 97%
Z-Trp-OBzl

Precaución: for Research Use Only

Formula: C₂₆H₂₄N₂O₄
MDL Number: MFCD00191163
CAS Number: 69876-37-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR