Ethyl-5-amino-1H-indole-2-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 5-Aminoindole-2-carboxylate

A569211-100MG A569211-250MG A569211-5G A569211-1G
76819-178EA 20 USD
76819-178 76819-180 77551-716 77884-390
Ethyl-5-amino-1H-indole-2-carboxylate 97%
Ethyl-5-amino-1H-indole-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₂N₂O₂
MW: 204.23 g/mol
MDL Number: MFCD08689973
CAS Number: 71086-99-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR