3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-5-(trifluoromethoxy)benzaldehyde 95+%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Formyl-5-(trifluoromethoxy)phenylboronic acid pinacol ester
A567111-100MG
A567111-1G
A567111-250MG
76819-170EA
33.95
USD
76819-170
76819-172
76819-174
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-5-(trifluoromethoxy)benzaldehyde 95+%
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-5-(trifluoromethoxy)benzaldehyde
Precaución: for Research Use Only
Formula:
C₁₄H₁₆BF₃O₄ MW: 316.08 g/mol |
MDL Number:
MFCD13181638 CAS Number: 1112209-48-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...