2,6-Difluoro-3-isopropoxyphenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (2,6-Difluoro-3-isopropoxyphenyl)boronic acid

A565006-1G A565006-250MG A565006-5G
76819-160EA 59 USD
76819-160 76819-162 76819-164
2,6-Difluoro-3-isopropoxyphenylboronic acid 98%
2,6-Difluoro-3-isopropoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BF₂O₃
MW: 215.99 g/mol
MDL Number: MFCD07369753
CAS Number: 849062-04-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR