4,4,5,5-Tetramethyl-2-(4-methyl-1-cyclohexen-1-yl)-1,3,2-dioxaborolane 98%

Supplier: Ambeed
Warning

A561932-100MG A561932-250MG A561932-1G A561932-5G
76819-152EA 31.83 USD
76819-152 76819-154 77594-708 77673-500
4,4,5,5-Tetramethyl-2-(4-methyl-1-cyclohexen-1-yl)-1,3,2-dioxaborolane 98%
4,4,5,5-Tetramethyl-2-(4-methyl-1-cyclohexen-1-yl)-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₃H₂₃BO₂
MW: 222.13 g/mol
MDL Number: MFCD11520541
CAS Number: 865869-26-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR