4-Bromo-2-iodo-6-nitroaniline 98%

Supplier: Ambeed
Warning

A560673-100MG A560673-1G A560673-250MG
76819-138EA 11.67 USD
76819-138 76819-140 76819-142
4-Bromo-2-iodo-6-nitroaniline 98%
4-Bromo-2-iodo-6-nitroaniline

Precaución: for Research Use Only

Formula: C₆H₄BrIN₂O₂
MW: 342.917 g/mol
MDL Number: MFCD11110560
CAS Number: 180624-08-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR