2-(2,6-Difluoro-3-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%

Supplier: Ambeed
Warning

Synonyms: 2,6-Difluoro-3-nitrophenylboronic acid pinacol ester

A558586-100MG A558586-1G A558586-250MG
76819-124EA 37 USD
76819-124 76819-126 76819-128
2-(2,6-Difluoro-3-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%
2-(2,6-Difluoro-3-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₂H₁₄BF₂NO₄
MDL Number: MFCD16295275
CAS Number: 1451391-10-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR