4-Carboxy-3-(trifluoromethyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-Borono-2-(trifluoromethyl)benzoic acid

A556916-1G A556916-250MG A556916-5G
76819-108EA 145.35 USD
76819-108 76819-110 76819-112
4-Carboxy-3-(trifluoromethyl)phenylboronic acid 98%
4-Carboxy-3-(trifluoromethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₆BF₃O₄
MW: 233.94 g/mol
MDL Number: MFCD11856032
CAS Number: 1050424-03-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR