Chelidamic acid monohydrate 98%

Supplier: Ambeed
Warning

Synonyms: 4-Oxo-1,4-dihydropyridine-2,6-dicarboxylic acid monohydrate

A556628-100G A556628-10G A556628-1G A556628-25G A556628-5G
76819-098EA 280.08 USD
76819-098 76819-100 76819-102 76819-104 76819-106
Chelidamic acid monohydrate 98%
Chelidamic acid monohydrate

Precaución: for Research Use Only

Formula: C₇H₅NO₅·H₂O
MW: 201.134 g/mol
MDL Number: MFCD00066478
CAS Number: 199926-39-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR