7-Amino-4,5-dihydro-5-oxopyrazolo[1,5-a]pyrimidine-3-carbonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 7-Amino-4,5-dihydro-5-oxo-pyrazolo[1,5-a]pyrimidine-3-carbonitrile

A555178-1G A555178-250MG
76819-094EA 155.96 USD
76819-094 76819-096
7-Amino-4,5-dihydro-5-oxopyrazolo[1,5-a]pyrimidine-3-carbonitrile 97%
7-Amino-4,5-dihydro-5-oxopyrazolo[1,5-a]pyrimidine-3-carbonitrile

Precaución: for Research Use Only

Formula: C₇H₅N₅O
MW: 175.15 g/mol
MDL Number: MFCD11040171
CAS Number: 138904-35-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR