1-Bromo-4-(tert-butoxy)benzene 97%

Supplier: Ambeed
Warning

A552805-10G A552805-1G A552805-25G A552805-5G
76819-066EA 55 USD
76819-066 76819-068 76819-070 76819-072
1-Bromo-4-(tert-butoxy)benzene 97%
1-Bromo-4-(tert-butoxy)benzene

Precaución: for Research Use Only

Formula: C₁₀H₁₃BrO
MDL Number: MFCD00792676
CAS Number: 60876-70-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR