2-Bromo-3-ethoxy-6-fluorophenylboronic acid 98%

Supplier: Ambeed
Warning

A552731-100MG A552731-1G A552731-250MG A552731-5G
76819-058EA 5 USD
76819-058 76819-060 76819-062 76819-064
2-Bromo-3-ethoxy-6-fluorophenylboronic acid 98%
2-Bromo-3-ethoxy-6-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₉BBrFO₃
MDL Number: MFCD06411368
CAS Number: 849052-19-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR