4-Chloro-3-(trifluoromethoxy)benzonitrile 98%

Supplier: Ambeed
Danger

A550587-1G A550587-5G
76819-054EA 166 USD
76819-054 76819-056
4-Chloro-3-(trifluoromethoxy)benzonitrile 98%
4-Chloro-3-(trifluoromethoxy)benzonitrile

Precaución: for Research Use Only

Formula: C₈H₃ClF₃NO
MDL Number: MFCD06660261
CAS Number: 886501-50-4
UN: 3439
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR