2-Chloro-3-iodobenzoic acid 95%

Supplier: Ambeed
Warning

A550222-100MG A550222-1G A550222-250MG
76819-046EA 21 USD
76819-046 76819-048 76819-050
2-Chloro-3-iodobenzoic acid 95%
2-Chloro-3-iodobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄ClIO₂
MDL Number: MFCD06808822
CAS Number: 874817-93-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR