6-Methylpyridine-3-sulfonyl chloride 95%

Supplier: Ambeed
Danger

A547422-100MG A547422-250MG A547422-5G A547422-1G
76819-032EA 35 USD
76819-032 76819-034 76819-036 77884-351
6-Methylpyridine-3-sulfonyl chloride 95%
6-Methylpyridine-3-sulfonyl chloride

Precaución: for Research Use Only

Formula: C₆H₆ClNO₂S
MDL Number: MFCD11868981
CAS Number: 478264-00-5
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR