3-Chloro-5-methylpyridazine 98%

Supplier: Ambeed
Warning

A547257-100MG A547257-1G
76819-024EA 23.34 USD
76819-024 76819-026
3-Chloro-5-methylpyridazine 98%
3-Chloro-5-methylpyridazine

Precaución: for Research Use Only

Formula: C₅H₅ClN₂
MDL Number: MFCD09743645
CAS Number: 89283-31-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR