1-Fluoro-4-iodo-2-methoxybenzene 97%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-5-iodoanisole

A546572-100MG A546572-1G A546572-250MG A546572-5G
76819-016EA 7 USD
76819-016 76819-018 76819-020 76819-022
1-Fluoro-4-iodo-2-methoxybenzene 97%
1-Fluoro-4-iodo-2-methoxybenzene

Precaución: for Research Use Only

Formula: C₇H₆FIO
MDL Number: MFCD09800754
CAS Number: 773855-64-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR