1-Boc-4-(4-Cyanobenzyl)piperazine 97%

Supplier: Ambeed
Warning

A545494-1G A545494-250MG A545494-5G
76819-008EA 46 USD
76819-008 76819-010 76819-012
1-Boc-4-(4-Cyanobenzyl)piperazine 97%
1-Boc-4-(4-Cyanobenzyl)piperazine

Precaución: for Research Use Only

Formula: C₁₇H₂₃N₃O₂
MDL Number: MFCD08460357
CAS Number: 849237-14-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR