Ticagrelor 98%

Supplier: Ambeed
Warning

A543807-10MG A543807-1MG A543807-250MG A543807-100MG A543807-1G A543807-5G
76818-994EA 6 USD
76818-994 76818-996 76818-998 77613-710 77613-712 77613-714
Ticagrelor 98%
Ticagrelor

Precaución: for Research Use Only

Formula: C₂₃H₂₈F₂N₆O₄S
MDL Number: MFCD09954148
CAS Number: 274693-27-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR