3-Chloropicolinic acid 98%

Supplier: Ambeed
Warning

A538028-100G A538028-10G A538028-250MG A538028-25G A538028-5G A538028-1G
76818-966EA 155 USD
76818-966 76818-968 76818-970 76818-972 76818-974 77594-656
3-Chloropicolinic acid 98%
3-Chloropicolinic acid

Precaución: for Research Use Only

Formula: C₆H₄ClNO₂
MDL Number: MFCD01646325
CAS Number: 57266-69-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR