(1S,2S)-Bis(4-methoxyphenyl)-1,2-ethanediamine 97%

Supplier: Ambeed
Warning

A537085-100MG A537085-1G A537085-250MG A537085-5G
76818-956EA 9 USD
76818-956 76818-958 76818-960 76818-962
(1S,2S)-Bis(4-methoxyphenyl)-1,2-ethanediamine 97%
(1S,2S)-Bis(4-methoxyphenyl)-1,2-ethanediamine

Precaución: for Research Use Only

Formula: C₁₆H₂₀N₂O₂
MDL Number: MFCD08460191
CAS Number: 58520-04-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR