4-Bromo-1-isopropylpyrazole 97%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-1-isopropyl-1H-pyrazole

A536564-1G A536564-250MG A536564-25G A536564-5G A536564-10G
76818-948EA 16.98 USD
76818-948 76818-950 76818-952 76818-954 77673-436
4-Bromo-1-isopropylpyrazole 97%
4-Bromo-1-isopropylpyrazole

Precaución: for Research Use Only

Formula: C₆H₉BrN₂
MW: 189.06 g/mol
MDL Number: MFCD09801021
CAS Number: 313735-62-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR