3-Chloro-1-methyl-1H-pyrazole 98%

Supplier: Ambeed
Warning

A534049-100MG A534049-1G A534049-5G A534049-250MG
76818-946EA 35 USD
76818-946 77569-034 77594-646 77673-430
3-Chloro-1-methyl-1H-pyrazole 98%
3-Chloro-1-methyl-1H-pyrazole

Precaución: For research use only.

Formula: C₄H₅ClN₂
MDL Number: MFCD09879994
CAS Number: 63425-54-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR