3-(Benzyloxy)-4-methoxybenzaldehyde 95%

Supplier: Ambeed
Warning

A533049-100G A533049-1G A533049-25G A533049-5G
76818-936EA 99 USD
76818-936 76818-938 76818-940 76818-942
3-(Benzyloxy)-4-methoxybenzaldehyde 95%
3-(Benzyloxy)-4-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₄O₃
MDL Number: MFCD00003386
CAS Number: 6346-05-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR