(4-Formyl-3,5-dimethylphenyl)boronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3,5-Dimethyl-4-formylphenylboronic acid

A532498-100MG A532498-1G A532498-250MG A532498-5G
76818-928EA 28 USD
76818-928 76818-930 76818-932 76818-934
(4-Formyl-3,5-dimethylphenyl)boronic acid 97%
(4-Formyl-3,5-dimethylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BO₃
MDL Number: MFCD15143475
CAS Number: 1218790-71-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR