2-Amino-2-(naphthalen-1-yl)ethanol 98%

Supplier: Ambeed
Warning

A528345-100MG A528345-250MG
76818-912EA 18 USD
76818-912 76818-914
2-Amino-2-(naphthalen-1-yl)ethanol 98%
2-Amino-2-(naphthalen-1-yl)ethanol

Precaución: for Research Use Only

Formula: C₁₂H₁₃NO
MDL Number: MFCD08752774
CAS Number: 86217-42-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR