4-(3-Pyridyloxy)benzenesulfonyl chloride hydrochloride 95%

Supplier: Ambeed
Danger

Synonyms: 4-(Pyridin-3-yloxy)benzene-1-sulphonyl chloride hydrochloride

A527431-100MG A527431-250MG
76818-908EA 63.66 USD
76818-908 76818-910
4-(3-Pyridyloxy)benzenesulfonyl chloride hydrochloride 95%
4-(3-Pyridyloxy)benzenesulfonyl chloride hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₈ClNO₃S·HCl
MW: 306.1700 g/mol
MDL Number: MFCD01862557
CAS Number: 1171901-60-2
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR