Methyl 4-amino-2-nitrobenzoate 98%

Supplier: Ambeed
Warning

A526900-1G A526900-250MG A526900-25G A526900-5G
76818-900EA 34 USD
76818-900 76818-902 76818-904 76818-906
Methyl 4-amino-2-nitrobenzoate 98%
Methyl 4-amino-2-nitrobenzoate

Precaución: for Research Use Only

Formula: C₈H₈N₂O₄
MDL Number: MFCD00228651
CAS Number: 84228-45-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR