Tetramethyl-1,3-cyclobutanedione 99%

Supplier: Ambeed
Warning

Synonyms: Tetramethylcyclobutane-1,3-dione

A522670-100G A522670-10G A522670-25G A522670-5G A522670-500G
76818-880EA 79.57 USD
76818-880 76818-882 76818-884 76818-886 77673-394
Tetramethyl-1,3-cyclobutanedione 99%
Tetramethyl-1,3-cyclobutanedione

Precaución: for Research Use Only

Formula: C₈H₁₂O₂
MW: 140.18 g/mol
MDL Number: MFCD00001331
CAS Number: 933-52-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR