3,4-Difluoro-2-methoxyphenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (3,4-Difluoro-2-methoxyphenyl)boronic acid

A520345-100G A520345-100MG A520345-25G
76818-866EA 705.5 USD
76818-866 76818-868 77613-638
3,4-Difluoro-2-methoxyphenylboronic acid 98%
3,4-Difluoro-2-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₇BF₂O₃
MW: 187.936 g/mol
MDL Number: MFCD13181640
CAS Number: 905583-06-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR