3-Bromo-5-chloro-4-hydroxybenzaldehyde 97%

Supplier: Ambeed
Danger

A515924-1G A515924-250MG A515924-5G
76818-826EA 43.5 USD
76818-826 76818-828 76818-830
3-Bromo-5-chloro-4-hydroxybenzaldehyde 97%
3-Bromo-5-chloro-4-hydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄BrClO₂
MW: 235.4625 g/mol
MDL Number: MFCD04129026
CAS Number: 1849-76-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR