5-Chloro-3H-spiro[isobenzofuran-1,4'-piperidine] hydrochloride 95%

Supplier: Ambeed
Warning

A514131-100MG A514131-1G A514131-250MG
76818-812EA 155.96 USD
76818-812 76818-814 76818-816
5-Chloro-3H-spiro[isobenzofuran-1,4'-piperidine] hydrochloride 95%
5-Chloro-3H-spiro[isobenzofuran-1,4'-piperidine] hydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₁₅Cl₂NO
MW: 260.16 g/mol
MDL Number: MFCD20921940
CAS Number: 1190965-20-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR