2-Bromo-3,3,3-trifluoropropan-1-ol 95%

Supplier: Ambeed
Danger

Synonyms: 1-Bromo-2-hydroxy-1-(trifluoromethyl)ethane, 2-Bromo-3-hydroxy-1,1,1-trifluoropropane

A511710-100G A511710-10G A511710-1G A511710-250MG A511710-25G A511710-5G
76818-794EA 2157.88 USD
76818-794 76818-796 76818-798 76818-800 76818-802 76818-804
2-Bromo-3,3,3-trifluoropropan-1-ol 95%
2-Bromo-3,3,3-trifluoropropan-1-ol

Precaución: for Research Use Only

Formula: C₃H₄BrF₃O
MW: 192.962 g/mol
MDL Number: MFCD09763638
CAS Number: 311-86-4
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR