3-Chloro-5-(trifluoromethyl)phenylboronic acid pinacol ester 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Chloro-5-(trifluoromethyl)benzeneboronic acid pinacol ester, 2-(3-Chloro-5-(trifluoromethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
A511660-1G
A511660-250MG
A511660-25G
A511660-5G
76818-786EA
47.75
USD
76818-786
76818-788
76818-790
76818-792
3-Chloro-5-(trifluoromethyl)phenylboronic acid pinacol ester 98%
3-Chloro-5-(trifluoromethyl)phenylboronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₃H₁₅BClF₃O₂ MW: 306.52 g/mol |
MDL Number:
MFCD11856039 CAS Number: 942069-65-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...