2-Fluoro-5-methoxypyridine 95%

Supplier: Ambeed
Warning

A511266-1G A511266-250MG A511266-5G
76818-784EA 35.01 USD
76818-784 77673-352 77673-354
2-Fluoro-5-methoxypyridine 95%
2-Fluoro-5-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₆FNO
MW: 127.116 g/mol
MDL Number: MFCD13185604
CAS Number: 136888-79-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR