3,4-Dibromofuran 96%

Supplier: Ambeed
Danger

A507012-100MG A507012-1G A507012-250MG A507012-5G
76818-772EA 9 USD
76818-772 76818-774 76818-776 77884-278
3,4-Dibromofuran 96%
3,4-Dibromofuran

Precaución: for Research Use Only

Formula: C₄H₂Br₂O
MDL Number: MFCD01318995
CAS Number: 32460-02-9
UN: 2922
ADR: 8,6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR