Methyl 2-(3-iodophenyl)acetate 98%

Supplier: Ambeed
Warning

A504606-1G A504606-250MG A504606-5G
76818-762EA 50 USD
76818-762 76818-764 77673-320
Methyl 2-(3-iodophenyl)acetate 98%
Methyl 2-(3-iodophenyl)acetate

Precaución: for Research Use Only

Formula: C₉H₉IO₂
MDL Number: MFCD13192188
CAS Number: 502649-73-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR