Potassium (2-cyanophenyl)trifluoroborate 95%

Supplier: Ambeed
Warning

A504361-1G A504361-25G A504361-5G
76818-756EA 51.99 USD
76818-756 76818-758 76818-760
Potassium (2-cyanophenyl)trifluoroborate 95%
Potassium (2-cyanophenyl)trifluoroborate

Precaución: for Research Use Only

Formula: C₇H₄BF₃N·K
MW: 209.018 g/mol
MDL Number: MFCD09952302
CAS Number: 929038-12-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR